missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Fraction V, 96%, Standard Grade, pH 5.2

For research use

Manufacturer:  Alfa Aesar™ J65855A1

 View more versions of this product

Catalog No. AAJ65855A1



Specifications

Specifications

Lyophilized Powder
5.2
1kg
2° to 8°C
Albumin, Bovine, Fraction V, 96%, Standard grade, pH 5.2
Lyophilized powder
Standard grade, 96%
Bovine Serum
RUO
No
Documents

For Research Use Only