missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Fraction V, 96%, Standard Grade, New Zealand origin, pH 7.0

For research use

Manufacturer:  Alfa Aesar™ J6578809

 View more versions of this product

Catalog No. AAJ6578809



Specifications

Specifications

Lyophilized Powder
7.0
10g
2° to 8°C
Albumin, Bovine, Fraction V, 96%, Standard grade, New Zealand origin, pH 7.0
Lyophilized powder
Standard grade, 96%
Bovine Serum
RUO
No
Documents

For Research Use Only