missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Fraction V, 96%, Standard Grade, New Zealand origin, pH 7.0

For research use

$84.38 - $1,448.95

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements 2° to 8°C
Source Bovine Serum
Product Type Albumin, Bovine, Fraction V, 96%, Standard grade, New Zealand origin, pH 7.0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6578809 View Documents Alfa Aesar™
J6578809
10g Each for $84.38
 
AAJ6578818 View Documents Alfa Aesar™
J6578818
50g Each for $342.09
 
AAJ6578822 View Documents Alfa Aesar™
J6578822
100g Each for $567.11
 
AAJ65788A1 View Documents Alfa Aesar™
J65788A1
1kg Each for $1,448.95
 
Specifications

Specifications

Lyophilized powder
2° to 8°C
Albumin, Bovine, Fraction V, 96%, Standard grade, New Zealand origin, pH 7.0
7.0
RUO
No
Bovine Serum
Lyophilized Powder
Standard grade, 96%

For Research Use Only