missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Reagent Grade, Fatty Acid Free, No Preservative

For research use

Manufacturer:  Alfa Aesar™ J65042K2

 View more versions of this product

Catalog No. AAJ65042K2



Specifications

Specifications

35% w/v
Liquid
Bovine Serum
RUO
No
Liquid
1L
2° to 8°C
Albumin, Bovine, Fraction V, 35% soln., Reagent grade, Fatty Acid Free, no preservative
Documents

For Research Use Only