missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Cell Culture Grade

For research use

Manufacturer:  Alfa Aesar™ J64101AE

 View more versions of this product

Catalog No. AAJ64101AE



Specifications

Specifications

35% w/v
Aqueous solution with 0.1% azide
Bovine Serum
RUO
No
Liquid
100mL
2° to 8°C
Albumin, Bovine, Fraction V, 35% soln., Cell Culture grade
Documents

For Research Use Only