Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67541LPL

 View more versions of this product

Catalog No. AAJ67541LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial
Product Type Albumin
Storage Requirements -30°C to -10°C
CAS 9048-46-8
Synonym Bovine Serum Albumim;
Format Lyophilized powder
Odor Odorless
Solubility Soluble in water at 100g/L
Description Pale green to white
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.