missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay Grade, Protease and Enzyme-free, pH5.2

For research use

Manufacturer:  Alfa Aesar™ J6597318

 View more versions of this product

Catalog No. AAJ6597318



Specifications

Specifications

Lyophilized Powder
5.2
50g
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay grade, protease and enzyme-free, pH5.2
Lyophilized powder
Immunoassay grade, 98%
Bovine Serum
RUO
No
Documents

For Research Use Only