missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay Grade, Protease and Enzyme-free, pH5.2

For research use

$156.59 - $1,871.61

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements 2° to 8°C
Source Bovine Serum
Product Type Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay grade, protease and enzyme-free, pH5.2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6597318 View Documents Alfa Aesar™
J6597318
50g Each for $156.59
 
AAJ6597336 View Documents Alfa Aesar™
J6597336
500g Each for $1,061.24
 
AAJ6597330 View Documents Alfa Aesar™
J6597330
250g Each for $661.38
 
AAJ65973A1 View Documents Alfa Aesar™
J65973A1
1kg Each for $1,871.61
 
Specifications

Specifications

Lyophilized powder
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay grade, protease and enzyme-free, pH5.2
RUO
5.2
No
Bovine Serum
Lyophilized Powder
Immunoassay grade, 98%

For Research Use Only