Please login to your online account to display your discounted pricing

Alfa Aesar™ Albendazole sulfone

$461.78 - $461.78

Chemical Identifiers

Chemical Name or Material Albendazole sulfone
CAS 75184-71-3
Molecular Formula C12H15N3O4S
Formula Weight 297.33
MDL Number MFCD00600775
Synonym Methyl 5-n-propylsulfonyl-2-benzimidazolecarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2591703

 
alfa aesar™
H2591703
1g Each for $461.78
Description & Specifications

Specifications

Formula Weight 297.33
MDL Number MFCD00600775
RTECS Number DD6520700
Chemical Name or Material Albendazole sulfone
Molecular Formula C12H15N3O4S
CAS 75184-71-3
Synonym Methyl 5-n-propylsulfonyl-2-benzimidazolecarbamate
Health Hazard 1 Warning
Melting Point 290° to 292°C
Quantity 1g
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).

Solubility
Soluble in DMSO, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.