missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Ala1,3,11,15] Endothelin-1, Human, Alfa Aesar™

For cellular and molecular biology applications

$147.05 - $233.04

Specifications

Product Type [Ala1,3,11,15] Endothelin-1, human
Cross Reactivity Human
Molecular Weight 2367.67
Storage Requirements -30° to -10°C
Sequence Ala-Ser-Ala-Ser-Ser-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66511EXD
View Documents
Thermo Fisher Scientific
J66511EXD
0.5mg
Each for $147.05
 
AAJ66511MCR
View Documents
Thermo Fisher Scientific
J66511MCR
1mg
Each for $233.04
 
Description

Description

Specifications

Specifications

[Ala1,3,11,15] Endothelin-1, human
2367.67
Ala-Ser-Ala-Ser-Ser-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
RUO
Human
-30° to -10°C
Solid
HPLC
SDS

For Research Use Only