Please login to your online account to display your discounted pricing

Alfa Aesar™ [Ala&superscript_9;,&superscript_1;&superscript_0;, Lys&superscript_1;&superscript_1;,&superscript_1;&superscript_2;] Glycogen Synthase (1-12)

For cellular and molecular biology applications

Specifications

Product Type [Ala9,10, Lys11,12] Glycogen Synthase (1-12)
Molecular Weight 1270.54
Storage Requirements -30° to -10°C
Sequence Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala-Lys-Lys
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66351LB0

 
alfa aesar™
J66351LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66351MCR

 
alfa aesar™
J66351MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Ala9,10, Lys11,12] Glycogen Synthase (1-12)
Molecular Weight 1270.54
Storage Requirements -30° to -10°C
Sequence Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala-Lys-Lys
Format Solid
Common Name PKC Substrate Peptide
Regulatory Status RUO
Purification Method HPLC