Please login to your online account to display your discounted pricing

Alfa Aesar™ [Ala&superscript_9;] Autocamtide 2

For cellular and molecular biology applications

Specifications

Product Type [Ala9] Autocamtide 2
Molecular Weight 1497.76
Storage Requirements -30° to -10°C
Sequence Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66658EXD

 
alfa aesar™
J66658EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66658MCR

 
alfa aesar™
J66658MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Ala9] Autocamtide 2
Molecular Weight 1497.76
Storage Requirements -30° to -10°C
Sequence Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu
Format Solid
Common Name Autocamtide-2-Related Inhibitory Peptide
Regulatory Status RUO
Purification Method HPLC