Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 8mm, Alfa Aesar™

$30.74 - $171.77

Chemical Identifiers

Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA32935DM

 
alfa aesar
32935DM
100mm Each for $30.74

AA32935DR

 
alfa aesar
32935DR
200mm Each for $49.86

AA32935DT

 
alfa aesar
32935DT
250mm Each for $59.38

AA32935DW

 
alfa aesar
32935DW
300mm Each for $68.86

AA32935EH

 
alfa aesar
32935EH
600mm Each for $105.47

AA32935EM

 
alfa aesar
32935EM
800mm Each for $136.50

AA32935EO

 
alfa aesar
32935EO
1000mm Each for $171.77
Description & Specifications

Specifications

Quantity 100mm
Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod