Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 5mm, Alfa Aesar™

$39.70 - $200.11

Chemical Identifiers

Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA32931DM

 
alfa aesar
32931DM
100mm Each for $39.70

AA32931DR

 
alfa aesar
32931DR
200mm Each for $59.57

AA32931DT

 
alfa aesar
32931DT
250mm Each for $75.88

AA32931DW

 
alfa aesar
32931DW
300mm Each for $77.59

AA32931EH

 
alfa aesar
32931EH
600mm Each for $129.92

AA32931EM

 
alfa aesar
32931EM
800mm Each for $161.62

AA32931EO

 
alfa aesar
32931EO
1000mm Each for $200.11

AA32931Q

 
alfa aesar
32931Q
400mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100mm
Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod