Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 4mm, Alfa Aesar™

$33.59 - $162.73

Chemical Identifiers

Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA32929DM

 
alfa aesar
32929DM
100mm Each for $33.59

AA32929DR

 
alfa aesar
32929DR
200mm Each for $48.97

AA32929DT

 
alfa aesar
32929DT
250mm Each for $59.51

AA32929DW

 
alfa aesar
32929DW
300mm Each for $61.03

AA32929EH

 
alfa aesar
32929EH
600mm Each for $108.18

AA32929EM

 
alfa aesar
32929EM
800mm Each for $132.45

AA32929EO

 
alfa aesar
32929EO
1000mm Each for $162.73
Description & Specifications

Specifications

Quantity 100mm
Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod