Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 3mm, Alfa Aesar™

$31.95 - $150.68

Chemical Identifiers

Chemical Name or Material Al-23 Solid Rod
Physical Form Solid Round Rod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA32927DM

 
alfa aesar
32927DM
100mm Each for $31.95

AA32927DR

 
alfa aesar
32927DR
200mm Each for $45.65

AA32927DT

 
alfa aesar
32927DT
250mm Each for $56.36

AA32927DW

 
alfa aesar
32927DW
300mm Each for $59.51

AA32927EM

 
alfa aesar
32927EM
800mm Each for $126.72

AA32927EH

 
alfa aesar
32927EH
600mm Each for $103.66

AA32927EO

 
alfa aesar
32927EO
1000mm Each for $150.68
Description & Specifications

Specifications

Quantity 100mm
Physical Form Solid Round Rod
Chemical Name or Material Al-23 Solid Rod