missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 2mm, Thermo Scientific™

$48.37 - $87.99

Specifications

Chemical Name or Material Al-23 Solid Rod
Form Rod
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA32926DM
View Documents Promotion Details
Thermo Scientific Chemicals
32926DM
100 mm
Each for $48.37
 
AA32926DR
View Documents Promotion Details
Thermo Scientific Chemicals
32926DR
200 mm
Each for $69.79
 
AA32926DT
View Documents Promotion Details
Thermo Scientific Chemicals
32926DT
250 mm
Each for $87.30
 
AA32926DW
View Documents Promotion Details
Thermo Scientific Chemicals
32926DW
300 mm
Each for $87.99
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Solid Rod
Rod
SDS
missing translation for 'documents'

missing translation for 'documents'