missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 10mm, Alfa Aesar™

$37.57 - $200.11

Chemical Identifiers

Physical Form Solid Round Rod
Chemical Name or Material Al-23 Solid Rod
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA32937DM Alfa Aesar
32937DM
100mm Each for $37.57
AA32937DR Alfa Aesar
32937DR
200mm Each for $53.26
AA32937DT Alfa Aesar
32937DT
250mm Each for $60.66
AA32937DW Alfa Aesar
32937DW
300mm Each for $71.43
AA32937EH Alfa Aesar
32937EH
600mm Each for $128.84
AA32937EM Alfa Aesar
32937EM
800mm Each for $160.08
AA32937EO Alfa Aesar
32937EO
1000mm Each for $200.11
Specifications Description & Specifications

Chemical Identifiers

Solid Round Rod
Al-23 Solid Rod

Specifications

100mm
Solid Round Rod
Al-23 Solid Rod