missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Al-23 Solid Rod, Round, Dia.: 1.5mm, Alfa Aesar™

$28.86 - $54.46

Chemical Identifiers

Physical Form Solid Round Rod
Chemical Name or Material Al-23 Solid Rod
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA32925DM Alfa Aesar
32925DM
100mm Each for $28.86
AA32925DR Alfa Aesar
32925DR
200mm Each for $43.82
AA32925DT Alfa Aesar
32925DT
250mm Each for $51.47
AA32925DW Alfa Aesar
32925DW
300mm Each for $54.46
Specifications Description & Specifications

Chemical Identifiers

Solid Round Rod
Al-23 Solid Rod

Specifications

100mm
Solid Round Rod
Al-23 Solid Rod