missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Al-23 Multibore Insulating Tube, O.D.: 8.5mm, Four 1.5mm Bores, Thermo Scientific™
$83.60 - $170.44
Specifications
Chemical Name or Material | Al-23 Multibore Insulating Tube |
---|---|
Name Note | Four bores |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA32560DV
|
Thermo Scientific Chemicals
32560DV |
275 mm |
Each for $83.60
|
|
|||||
AA32560DZ
|
Thermo Scientific Chemicals
32560DZ |
380 mm |
Each for $106.64
|
|
|||||
AA32560EL
|
Thermo Scientific Chemicals
32560EL |
770 mm |
Each for $170.44
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Al-23 Multibore Insulating Tube | |
Four bores |