Please login to your online account to display your discounted pricing

Al-23 Insulating Beads, O.D.: 4mm, Four 0.8mm Bores, Length: 4mm, Alfa Aesar™

$140.08 - $140.08

Chemical Identifiers

Chemical Name or Material Al-23 Insulating Beads
Name Note 4 channels
Physical Form Insulating Beads
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA32914KX

 
alfa aesar
32914KX
100 beads Each for $140.08
Description & Specifications

Specifications

Physical Form Insulating Beads
Quantity 100 beads
Chemical Name or Material Al-23 Insulating Beads
Name Note 4 channels