Please login to your online account to display your discounted pricing

Al-23 Grinding Pieces, H: 20mm, 53 Pieces/Kg, Alfa Aesar™

$289.30 - $8,770.85

Chemical Identifiers

Chemical Name or Material Al-23 Grinding Pieces
Physical Form Grinding Pieces
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA33192A1

 
alfa aesar
33192A1
1kg Each for $289.30

AA33192A7

 
alfa aesar
33192A7
5kg (5 x 53 Pieces) Each for $1,137.61

AA33192A9

 
alfa aesar
33192A9
10kg Each for $2,034.12

AA33192C6

 
alfa aesar
33192C6
50kg (50 x 53 Pieces) Each for $8,770.85
Description & Specifications

Specifications

Quantity 1kg
Physical Form Grinding Pieces
Chemical Name or Material Al-23 Grinding Pieces