Please login to your online account to display your discounted pricing

Al-23 Combustion Tray, L1: 50mm, L2: 48mm, H: 20mm, W: 3mm, Vol.: 33, Alfa Aesar™

$138.54 - $138.54

Chemical Identifiers

Chemical Name or Material Al-23 Combustion Tray
Physical Form Tray
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA33184KF

 
alfa aesar
33184KF
1 tray Each for $138.54
Description & Specifications

Specifications

Quantity 1 tray
Chemical Name or Material Al-23 Combustion Tray
Physical Form Tray