AKT3 (PKB gamma) Recombinant Human Protein DFS Item
AKT3 (PKB gamma)
Manufacturer:  Life Technologies LSPV3976
 View more versions of this product
Catalog No. PV3976
Description
Description
AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). It is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake.
Specifications
Specifications
NP_005456
Protein assays and analysis
Frozen
AKT3 (PKB gamma) Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
AKT3 (PKB gamma)
AKT3 (PKB gamma) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM Na3VO4, 1mM DTT, 0.01% Triton™ X-100, 200μM ATP, 50μM crosstide peptide substrate (GRPRTSSFAEG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
AKT3 (PKB gamma) Recombinant Human Protein
Human
0.48mg/mL
Frozen
60.4kDa
95% by SDS-PAGE
–80°C
1 Each
AKT3 (PKB gamma) Recombinant Human Protein
Kinase assay
AKT
300nmoles of phosphate transferred to Crosstide peptide substrate (GRPRTSSFAEG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.