missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ AKT2 (PKB beta) Recombinant Human Protein
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues.
$2650.00 - $11260.00
Specifications
Accession Number | NP_001617 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol |
Molecular Weight (g/mol) | 59.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR4640B
|
Invitrogen™
PR4640B |
100μg |
Each for $2,650.00
|
|
|||||
PV3975
|
Invitrogen™
PV3975 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
AKT2 has been implicated in the metabolic actions of the hormone.
Specifications
NP_001617 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
AKT Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
59.1kDa | |
RUO - research use only | |
AKT2 (PKB beta) | |
Human | |
His Tag | |
Frozen |