Invitrogen™ AKT2 (PKB beta) Recombinant Human Protein DFS Item
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues.
Manufacturer:  Invitrogen™ LSPV3975
 View more versions of this product
Catalog No. PV3975
Description
Description
AKT2 has been implicated in the metabolic actions of the hormone.
Specifications
Specifications
NP_001617
Frozen
59.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
AKT Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol
1mg
RUO - research use only
AKT2 (PKB beta)
Baculovirus
Human
His Tag
Documents