missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Akro-Mils™ InSight™ Storage System: 15 lb., 10 7/8 x 4 1/8 x 3 1/4 in.

Visible organization solutions for healthcare and industrial workspaces

Manufacturer:  Fisher Scientific 305A5

 View more versions of this product

Catalog No. 08757643


 

Specifications Description & Specifications

Specifications

Storage Bin
15 lb.
3.12 in.
23.4cm
6.08kg
Polycarbonate
8.25cm
10.8 x 4.37 x 3.25 in. (27.4 x 11 x 8.25cm)
3.25 in.
11cm
4.37 in.
InSight Storage System
10.8 in.
6.65cm
2.62 in.
27.4cm
8cm
9.25 in.
Yes