Please login to your online account to display your discounted pricing

Akro-Mils™ InSight™ Storage System: 10 lb., 5 3/8 x 4/18 x 3 1/4 in.

Visible organization solutions for healthcare and industrial workspaces

$25.27 - $203.13

Specifications

Autoclavable Yes
Material Polycarbonate
For Use With InSight Storage System
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

08757641

 
fisher scientific
305A1
Storage Bin Case of 16 Each for $203.13

08757670

 
fisher scientific
405A1
Length divider Pack of 8 Each for $25.27

08757674

 
fisher scientific
305A2
Storage bin lid Case of 16 Each for $165.87
Description & Specifications

Specifications

Autoclavable Yes
Material Polycarbonate
For Use With InSight Storage System

InSight bins offer transparent storage with a sleek, modern design. Can be stacked, shelved or hung on Akro-Mils rails and louvered panels or ReadySpace™ Storage System.

  • Constructed of durable polycarbonate
  • Autoclavable
  • No tip feature for secure stacking
  • Rear lip supports bin on hanging systems
  • Large label area for easy identification and bar code scanning
  • Optional lids and length/width dividers available to create dust-free, custom-size compartments