Please login to your online account to display your discounted pricing

Akro-Mils™ AkroBins™ Small Storage Bin Accessories: Bin Dividers


Bin Dividers for use with Akro-Mils AkroBins Small Storage Bins

Manufacturer: fisher scientific  40220

 View more versions of this product

Catalog No. 08757612

  • / Pack of 24 Each
 

Description & Specifications

Specifications

Product Type Bin Dividers for Akro-Mils AkroBins Small Storage Bins
For Use With AkroBins (08-757-624)
Color Black
Dimensions (L x W x H) 7.38L x 4.12W x 3 in. H