missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Akro-Mils™ AkroBins™ Large Storage Bins

Molded of high-impact polypropylene

Manufacturer:  Akro-Mils 30240SCLAR

 View more versions of this product

Catalog No. 08-757-718


 

Specifications Description & Specifications

Specifications

14.75 x 8.25 x 7 in. (375 x 210 x 178mm)
Polypropylene
7 in.
14.75 in.
8.25 in.
Clear
14 in.
Storage bin
60 lb.
178mm
375mm
210mm
6.75 in.
6.56 in.