missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Akro-Mils™ AkroBins™ Large Storage Bins

Molded of high-impact polypropylene

Manufacturer:  Akro-Mils 30260RED

 View more versions of this product

Catalog No. 08-757-731


 

Specifications Description & Specifications

Specifications

18 x 11 x 10 in. (457 x 279 x 254mm)
Polypropylene
10 in.
18 in.
11 in.
Red
17 in.
Storage bin
60 lb.
254mm
457mm
279mm
9.75 in.
9.5 in.