Please login to your online account to display your discounted pricing

Akro-Mils™ AkroBins™ Extra Large Storage Bins: Dividers

For use with extra large AkroBins

$78.50 - $86.77

Specifications

Color Black
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W x H) For Use With Price Quantity    

08757626

 
fisher scientific
40270
18 x 16.5 x 11 in. (457 x 420 x 279mm) 18 x 16.5 x 11 in. Bins Pack of 6 Each for $86.77

08757636

 
fisher scientific
40265*
18 x 8.25 x 9 in. (457 x 209 x 229mm) 18 x 8.25 x 9 in. Bins Pack of 6 Each for $78.50
Description & Specifications

Specifications

Color Black
Material Polypropylene

  • Molded of high-impact polypropylene
  • Unaffected by weak acids and alkalis
  • Create customizable compartments