Please login to your online account to display your discounted pricing

(+)-AJ 76 hydrochloride, Tocris

Antagonist; preferential action at D2-like autoreceptors

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material (+)-AJ 76 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H23NO·HCl
Formula Weight 269.81
Synonym (1S, 2R)-cis-5-Methoxy-1-methyl-2-(N-propylamino)tetralin hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

067810

 
r & d systems
RDS067810
10mg Each for $199.70

067850

 
r & d systems
RDS067850
50mg Each for $834.54
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 269.81
Chemical Name or Material (+)-AJ 76 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H23NO·HCl
CAS 85378-82-1
Synonym (1S, 2R)-cis-5-Methoxy-1-methyl-2-(N-propylamino)tetralin hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature