Please login to your online account to display your discounted pricing

(+)-AJ 76 hydrochloride, Tocris


Antagonist; preferential action at D2-like autoreceptors

Quantity 50mg

Manufacturer: r & d systems  RDS067850

 View more versions of this product

Catalog No. 067850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 269.81
CAS 85378-82-1
Chemical Name or Material (+)-AJ 76 hydrochloride
Synonym (1S, 2R)-cis-5-Methoxy-1-methyl-2-(N-propylamino)tetralin hydrochloride
Assay Percent Range >98%
Molecular Formula C15H23NO·HCl
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (+)-AJ 76 hydrochloride
Assay Percent Range >98%
Molecular Formula C15H23NO·HCl
Formula Weight 269.81
Synonym (1S, 2R)-cis-5-Methoxy-1-methyl-2-(N-propylamino)tetralin hydrochloride
Accessories

Store at Room Temperature