Please login to your online account to display your discounted pricing

AIRE Rat anti-Human, Clone: TM-724, eBioscience™

Rat Monoclonal Antibody

$160.95 - $265.35

Specifications

Antigen AIRE
Clone TM-724
Host Species Rat
Gene Alias Autoimmune Regulator
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122791

 
life technologies
LS14953480
25μg Each for $160.95

501122792

 
life technologies
LS14953482
100μg Each for $265.35
Description & Specifications

Specifications

Antigen AIRE
Clone TM-724
Host Species Rat
Gene Alias Autoimmune Regulator
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This TM-724 monoclonal antibody reacts with human Autoimmune Regulator (AIRE). This 58kDa transcription factor is highly expressed within the perinuclear region of mature medullary thymic epithelial cells (mTECs). AIRE mRNA in the periphery has been observed in the lymph node, fetal liver, and intestines. Furthermore expression has been observed in dendritic cells, epithelial cells and PBMCs. Studies demonstrate that AIRE is involved in thymocyte development, in particular the regulation of tissue-specific antigen expression during negative selection. Other biological functions for AIRE include chemokine expression and antigen presentation. As a result of alternative splicing, three isoforms have been reported. The conditions autoimmune–polyendocrinopathy–candiasis–ectodermal–dystrophy (APECED) and autoimmune polyglandular syndrome type 1 (APS-1) arise from mutations in the human AIRE gene.

The TM-724 antibody is predicted to recognize all three isoforms of human AIRE (AIRE-1, AIRE-2, AIRE-3).