AirClean™ Systems PowderSafe™ Type B Enclosure Accessory, Prefilter
AirClean™ Systems PowderSafe™ Type B Enclosure Accessory, Prefilter is an electrostatically charged fiber filter that removes sub-micron particulates.
Manufacturer: AirClean™ Systems ACFPRE8
Description
- Electrostatically charged fiber filter that removes sub-micron particulates
- Has efficiency rating of 99.997% at 0.3 microns
- Used to protect absorption filters from contamination
- Each absorption filter is fitted with a pre-filter at time of purchase
Specifications
Prefilter | |
PowderSafe Type B Enclosure |