AirClean™ Systems HEPA and ULPA Filters
Manufacturer:  AirClean™ Systems ACFHEPADUP*
Catalog No. 36-099-3879
Description
Description
  • Electrostatically charged filters prevent “particle-rain“
  • Metal screened on face of filter to promote even air distribution and laminar airflow
  • Metal-free filters available upon request