AirClean™ Systems HEPA and ULPA Filters
For removal of powders, fibers and particulate
$617.86 - $617.86
Specifications
Material Activated Carbon
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Efficiency Filter Type Price Quantity  
Catalog Number Mfr. No. Efficiency Filter Type Price Quantity  
36-099-3879 AirClean™ Systems
ACFHEPA
99.997% ACFHEPA Each for $617.86
Specifications
Description
  • Electrostatically charged filters prevent “particle-rain“
  • Metal screened on face of filter to promote even air distribution and laminar airflow
  • Metal-free filters available upon request
Description & Specifications
Specifications
Activated Carbon