AirClean™ Systems Bulk Powder Handling Enclosures
Provide effective control and containment of airborne particulate during manipulation and transfer of potent compounds
$29905.61 - $38845.72
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
36-101-8900
|
AirClean™ Systems
AC794BE |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
36-101-8901
|
AirClean™ Systems
AC795BE |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
36-101-8902
|
AirClean™ Systems
AC796BE |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Designed to provide turbulent free airflow environment
- Air moves in horizontal pattern to maximize containment while minimizing sample loss and balance instability
- HEPASafe™ filter change-out technology allows operator to safely and effectively change both pre-filter and primary HEPA filter
- Preset face velocity that is constantly monitored and maintained by AirSafe™ automatic safety controller
- Audible and visible alarms when filter change out is necessary
- Optional bonded carbon filtration for removal of fumes during bulk chemical handling and dispensing is also available
- With sturdy stand which provides comfortable working height of 39 in.
- Available in 4 ft., 5 ft. and 6 ft. widths with nominal depth of 36 in.
Specifications
Bulk Powder Handling Enclosure |