AirClean™ Systems PowderSafe Type B Enclosures
Choose the Airclean PowderSafe Type B Enclosure for powder weighing and containment technology.
Manufacturer:  AirClean™ Systems AC730C
 View more versions of this product
Catalog No. 36-100-4292
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • AirSafe™ automatic safety controller
  • Solid polypropylene construction
  • Effectively weigh to 5 decimal places
  • Electrical cord access ports
  • Continuous HEPA filter monitoring
  • HEPASafe™ filter change technology provides a safe and effective means for replacement of both the pre-filter and primary HEPA filter
  • Audible and visible alarms
  • Real Time airflow display on LCD
  • Portable
  • Fluorescent light with one touch operation
  • Modular electronics for easy service
  • Optional sturdy mobile cart
Specifications
Specifications
33 in.
HEPA
18 in.
45.72cm
32 in.
Filter change technology allows for safe and easy change of all contaminated filters, while under negative pressure
83.82cm
53.3 x 78.7 x 45.7cm (21 x 31 x 18 in.)