AirClean™ Systems Polypropylene Horizontal Laminar Flow Clean Bench
For Class 100 (ISO 5) applications
Manufacturer:  AirClean™ Systems AC8000HLFDUP*
Catalog No. 36-100-4285
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • UVTech™ microprocessor controller constantly monitors filter conditions
  • Polypropylene construction
  • Seamless, smooth construction minimizes air turbulence
  • Built-in fluorescent lighting
  • All-white surfaces are easy to clean
  • Available in 110/220V versions
  • Supplied fully assembled
Options
  • Custom sturdy carts or stands
  • Cup sink and vacuum fittings
  • Polypropylene base cabinet
  • Ultraviolet light source
  • IV bar
This product will not protect the operator from biological hazards. Do not use this product with biological hazardous material.
  • IV admixture preparation
  • Drug compounding
  • Plant cell culture
  • Tissue culture (non-biological)
  • Media preparation
  • Pharmaceutical procedures
  • Electronic assembly
Certifications
UL listed, CSA Certified
Compliance
Meets ISO 5 and FS209E Class 100 clean bench standards.