Please login to your online account to display your discounted pricing
Airclean™ System AC600 Series Ductless Endoscopy Workstations
Provide mobile solution for operator protection from high-level disinfectant vapors during endoscope reprocessing
$5,415.97 - $5,415.97
Specifications
Certifications/Compliance ANSI Z9.5, A.A.M.I. ST58, OSHA and JCAHO
Filter Glutaraldehyde, OPA, Cidex™, Peracetic Acid, Hydrogen Peroxide
For Use With (Application) A mobile solution for operator protection from disinfectant fumes
Height (English) Interior 22 in.
View More Specs
Products
Catalog Number Mfr. No. Airflow Height (Metric) Exterior Height (English) Exterior Width (English) Exterior Price Quantity    
36-100-0007
airclean systems
AC632C
200 cfm 76.2cm 30 in. 32 in. Each for $5,415.97
Description & Specifications
Specifications
Certifications/Compliance ANSI Z9.5, A.A.M.I. ST58, OSHA and JCAHO
Filter Glutaraldehyde, OPA, Cidex™, Peracetic Acid, Hydrogen Peroxide
For Use With (Application) A mobile solution for operator protection from disinfectant fumes
Height (English) Interior 22 in.
  • AirSafe™ automatic safety controller for complete system monitoring
  • Chemisorptive bonded carbon neutralizing filter
  • Accommodates virtually any layout of disinfectant and rinse trays
  • For use with rigid or flexible endoscopes
  • No installation or ducting required
  • Integral base holds up to 3 gal. spill
  • Bi-fold sash provides easy access to trays and acts as eye shield against potential splashing
  • Filter and electronics are located above work area, preventing spills from contacting these components
Options:
  • GlutAway™ waste pump for transferring disinfectant and rinse water to waste
  • Sturdy cart with shelf
ANSI Z9.5, OSHA and JCAHO requirements, A.A.M.I. ST58