Air Track Accessories
Use with air track (S52229) to study linear motion.
Manufacturer:  Fisher Science Education 12514
 View more versions of this product
Catalog No. S52204C
Specifications Description & Specifications
Specifications
Air track (S52229)
Each
7.5cm
Glider
3 in.
Four 105g masses, screws, wingnuts, bumpers, threaded center hole