missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Air Systems™ Venturi-Style Pneumatic Air Blowers

$497.22 - $497.22

Specifications

Diameter (English) Fan 7.37 in.
Flow Rate 38cfm at 50PSIG
Length (English) 32.12 in.
Length (Metric) 81.58cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
18-502 Air Systems International
ASI1200
Venturi pneumatic air blower Each for $497.22
Specifications

Description

Aluminum base and galvanized steel horn. Expel explosive and hazardous vapors or fumes from underground spaces. No moving parts—horns operate on compressed air or steam. Horn 18-503 meets ventilation requirements for underground storage tanks. Static grounding lug is standard on all models. Horns stand on lightweight aluminum base.

Description & Specifications

Specifications

7.37 in.
38cfm at 50PSIG
32.12 in.
81.58cm