Air Systems™ Auto-Air™ Breather Box™ Air Filtration System Accessory, Calibration Kit
Kits for use with Carbon Monoxide (CO) monitor.
Supplier: Air Systems™ BBK20
Description
For use with Air Systems Carbon Monoxide Monitor (Cat. No. 19-806-351)
Specifications
Carbon Monixide Monitor | |
BBK-20 | |
Calibration Kit |
2 pressurized 17 L disposable cylinders -one cylinder contains zero air, the other contains 20 ppm of CO air, flow regulator, pressure gauge, vinyl tubing in hard carry case | |
Small |