Air Source
Compact, one-horsepower unit provides enough air for two air tracks.
$56.40 - $56.40
Specifications
Quantity Each
For Use With (Application) To operate basic precision air track and most other air tracks
View More Specs
Includes: Hose, 6.6 ft. (2m) long
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity  
S522277 Fisher Science Education
EA21
For air source Spare air hose Each for $56.40
Specifications
Description
  • Two outlets offer simultaneous operation of two air tracks
  • Air intake absorber minimizes noise while providing a straight-through air path for optimum compressor cooling
  • Designed to operate with the Basic Precision Air Track (S52227ND) and most other air tracks
Order Info
Dimensions: 11.75L × 6.25W × 6.75 in. H; weight: 6.75 lb. (3.1kg)
Description & Specifications
Specifications
Each
To operate basic precision air track and most other air tracks