missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aiolos Mouse anti-Mouse, eFluor 660, Clone: 8B2, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $473.88

Specifications

Antigen Aiolos
Clone 8B2
Host Species Murine
Gene Alias IKZF3
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124398 Life Technologies
LS50578980
25μg Each for $183.48
501124399 Life Technologies
LS50578982
100μg Each for $473.88
Specifications

Description

This 8B2 monoclonal antibody reacts with mouse Aiolos, a 58kDa member of the Ikaros family of zinc finger transcription factors. Expression of Aiolos is restricted to the lymphoid lineage, with B cells exhibiting the highest levels of expression, followed by NK and T cells. Studies demonstrate that Aiolos forms homodimers or heterodimerizes with Ikaros proteins. Moreover, Aiolos exhibits tyrosine phosphorylation and nuclear localization in response to IL-2 stimulation in T cells. Aiolos plays a role in IL-2-mediated anti-apoptosis by controlling Bcl-2 expression. Finally, Aiolos is involved in B cell development, with upregulation of this transcription factor observed at the pre-BI to pre-BII transition in response to pre-B cell receptor signaling.
Description & Specifications

Specifications

Aiolos
Murine
Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
8B2
IKZF3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer