Please login to your online account to display your discounted pricing

AIDA, Tocris


Potent, selective group I mGlu antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS090410

 View more versions of this product

Catalog No. 090410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 221.21
CAS 168560-79-0
Synonym (RS)-1-Aminoindan-1, 5-dicarboxylic acid
Assay Percent Range >98%
Chemical Name or Material AIDA
Molecular Formula C11H11NO4
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material AIDA
Assay Percent Range >98%
Molecular Formula C11H11NO4
Formula Weight 221.21
Synonym (RS)-1-Aminoindan-1, 5-dicarboxylic acid
Accessories

Store at Room Temperature