Please login to your online account to display your discounted pricing

AH 6809, Tocris


EP1 and EP2 receptor antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS067150

 View more versions of this product

Catalog No. 067150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 298.3
Chemical Name or Material AH 6809
Assay Percent Range >98%
Molecular Formula C17H14O5
CAS 33458-93-4
Synonym 6-Isopropoxy-9-xanthone-2-carboxylic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material AH 6809
Assay Percent Range >98%
Molecular Formula C17H14O5
Formula Weight 298.3
Synonym 6-Isopropoxy-9-xanthone-2-carboxylic acid
Accessories

Store at Room Temperature