Please login to your online account to display your discounted pricing

AH 11110 hydrochloride, Tocris


Subtype-selective α1B ligand

Quantity 50mg

Manufacturer: r & d systems  RDS099850

 View more versions of this product

Catalog No. 099850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 374.91
Chemical Name or Material AH 11110 hydrochloride
Assay Percent Range >98%
Molecular Formula C21H26N2O2·HCl
CAS 179388-65-9
Synonym 4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material AH 11110 hydrochloride
Assay Percent Range >98%
Molecular Formula C21H26N2O2·HCl
Formula Weight 374.91
Synonym 4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
Accessories

Store at Room Temperature